Learn More
Apexbio Technology LLC Dextrose (D-glucose) 50-99-7 5g
SDP
Catalog No. 5000096213
Encompass_Preferred
Dextrose (D-glucose) (CAS 50-99-7) is a monosaccharide serving as an endogenous ligand for glucose transporters (GLUTs) It facilitates cellular glucose uptake and ATP generation by acting as a primary metabolic substrate Dextrose exerts its biological activity chiefly through glycolytic metabolism supporting cell growth proliferation and survival In cell-based studies it is widely applied as a metabolic substrate in cell culture media and as glucose supplementation during cell viability assays metabolic assessments and as a reference agent in glucose uptake and insulin sensitivity investigations Dextrose itself generally lacks a defined IC50 value due to its role as a natural substrate Based on these pharmacological properties Dextrose is utilized in research on cellular metabolism viability and insulin responsiveness
Catalog No. 50-000-96213 Supplier Apexbio Technology LLC Supplier No. A84065000
Dextrose (D-glucose) (CAS 50-99-7) is a monosaccharide serving as an endogenous ligand for glucose transporters (GLUTs) It facilitates cellular glucose uptake and ATP generation by acting as a primary metabolic substrate Dextrose exerts its biological activity chiefly through glycolytic metabolism supporting cell growth proliferation and survival In cell-based studies it is widely applied as a metabolic substrate in cell culture media and as glucose supplementation during cell viability assays metabolic assessments and as a reference agent in glucose uptake and insulin sensitivity investigations Dextrose itself generally lacks a defined IC50 value due to its role as a natural substrate Based on these pharmacological properties Dextrose is utilized in research on cellular metabolism viability and insulin responsiveness