Learn More
MOLTOX Molecular Toxicology Inc Dextrose Starch Agar, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948819
Encompass_Preferred
Dextrose Starch Agar is used for cultivating fastidious microorganisms, primarily Neisseria gonorrhoea as a pure culture. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-819 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S021
Dextrose Starch Agar is used for cultivating fastidious microorganisms, primarily Neisseria gonorrhoea as a pure culture. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.