Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextrose Starch Agar | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500 G
Catalog No. NC2680693
Encompass
Dextrose Starch Agar | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500gThis bacteriological growth medium contains dextrose, soluble starch, peptone, sodium chloride and agar. It is used for the isolation and differentiation of microorganisms based on their ability to produce starch-hydrolyzing enzymes. Dextrose and soluble starch provide carbon and energy sources. Peptone supplies nitrogen. Sodium chloride maintains osmotic equilibrium. Agar acts as the gelling agent.
Catalog No. NC2680693 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D3060500G
Dextrose Starch Agar | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500gThis bacteriological growth medium contains dextrose, soluble starch, peptone, sodium chloride and agar. It is used for the isolation and differentiation of microorganisms based on their ability to produce starch-hydrolyzing enzymes. Dextrose and soluble starch provide carbon and energy sources. Peptone supplies nitrogen. Sodium chloride maintains osmotic equilibrium. Agar acts as the gelling agent.