Learn More
Bioworld Dextrose Tryptone Agar, 500 g
Catalog No. 50254299
Encompass_Preferred
Dextrose Tryptone AgarUsed for the cultivation of "FLAT SOUR" organisms related to the spoilage of food productsIs a non-selective medium which can support the growth of a variety of mesophilic and thermophilic microorganismspH: 6.7Working Concentration: 30 g/LDehydrated Appearance: light green-beige, homogeneous powderPrepared Appearance: purple agarSoluble in boiling waterComposition:Tryptone Dextrose Bromcresol Purple Agar
Catalog No. 50-254-299 Supplier Bioworld Supplier No. 306200221
Dextrose Tryptone AgarUsed for the cultivation of "FLAT SOUR" organisms related to the spoilage of food productsIs a non-selective medium which can support the growth of a variety of mesophilic and thermophilic microorganismspH: 6.7Working Concentration: 30 g/LDehydrated Appearance: light green-beige, homogeneous powderPrepared Appearance: purple agarSoluble in boiling waterComposition:Tryptone Dextrose Bromcresol Purple Agar