Learn More
Bioworld Dextrose Tryptone Broth, 500 g
Supplier:  Bioworld 306200231
Encompass_Preferred
Dextrose Tryptone BrothUsed for the cultivation of "FLAT SOUR" organisms related to the spoilage of food productsIs a non-selective medium which can support the growth of a variety of mesophilic and thermophilic microorganismspH: 6.7Working Concentration: 30 g/LDehydrated Appearance: light green-beige, homogeneous powderPrepared Appearance: dark burghuny brothSoluble in waterComposition:Tryptone Dextrose Bromcresol Purple
Catalog No. 50-254-300