Learn More
NEOGEN Dextrose Tryptone Broth, 5 kg
SDP
Catalog No. 502015367
Encompass_Preferred
Dextrose Tryptone Broth is used for cultivation of mesophilic or thermophilic spoilage microorganisms from food in a laboratory setting. Dextrose Tryptone Broth is not intended for use in the diagnosis of disease or other conditions in humans.In the 1930’s, the National Canners Association specified use of Dextrose Tryptone Agar for isolating “flat sour” organisms from food products. “Flat sour” spoilage of canned foods is caused by Bacillus coagulans (Bacillus thermoacidurans). Bacterial growth results in a 0.3 – 0.5 drop in pH, while ends of the can remain flat. B. coagulans is a soil microorganism and found in canned tomato products and dairy products. Conditions favorable for multiplication of the bacterium can result in spoilage of the food product.For more information visit https://www.neogen.com/solutions/Microbiology/dextrose-tryptone-broth
Catalog No. 50-201-5367 Supplier NEOGEN Supplier No. 700003174
Dextrose Tryptone Broth is used for cultivation of mesophilic or thermophilic spoilage microorganisms from food in a laboratory setting. Dextrose Tryptone Broth is not intended for use in the diagnosis of disease or other conditions in humans.In the 1930’s, the National Canners Association specified use of Dextrose Tryptone Agar for isolating “flat sour” organisms from food products. “Flat sour” spoilage of canned foods is caused by Bacillus coagulans (Bacillus thermoacidurans). Bacterial growth results in a 0.3 – 0.5 drop in pH, while ends of the can remain flat. B. coagulans is a soil microorganism and found in canned tomato products and dairy products. Conditions favorable for multiplication of the bacterium can result in spoilage of the food product.For more information visit https://www.neogen.com/solutions/Microbiology/dextrose-tryptone-broth