Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Dextrose USP (Anhydrous)
SDP

Catalog No. 14186471 Shop All Medisca Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-471 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-186-471 Supplier Medisca Supplier No. 007201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextrose USP comes as colorless crystals or white, crystalline or granular powder. It is odorless. Alternative name(s): D-Glucose, glucose

Dextrose USP comes as colorless crystals or white, crystalline or granular powder. It is odorless.

Alternative name(s): D-Glucose, glucose

Specifications

Chemical Name or Material Dextrose USP (Anhydrous)
Packaging Jar, 4L, White, HDPE, Round, 110-400,TE Pano Neck
CAS 50-99-7
Type Drug Standard
Molecular Weight (g/mol) 180.16
Quantity 2.5 kg
Percent Purity 97.5 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.