Learn More
Abcam Carbo x yl-DCFDA (5(6)-Carbo x y-2',7'-dichlorofluorescein diacetate), Fluorogenic esterase substrate, 100MG
Catalog No. 502705214
Encompass_Preferred
MW 529.28 Da, Purity >85%. Amine-reactive, fluorogenic, non-selective esterase substrate. Hydrolyses to DFCA (ab145439) fluorophore intracellularly. Labels intact cells. 2,7-Dichlorofluorescein (ab146332) derivative and DCFDA N-succinimidyl ester (ab145286) precursor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5214 Supplier Abcam Supplier No. AB145415100MG
MW 529.28 Da, Purity >85%. Amine-reactive, fluorogenic, non-selective esterase substrate. Hydrolyses to DFCA (ab145439) fluorophore intracellularly. Labels intact cells. 2,7-Dichlorofluorescein (ab146332) derivative and DCFDA N-succinimidyl ester (ab145286) precursor.
The product is subject to the following: Abcam Restricted Use Statement