Learn More
Medchemexpress LLC Dfdbi-2T | 1539318-40-5 | 98.0% | 306.19 | 1 ML
SDP
Catalog No. 5000344013
Encompass_Preferred
DFHBI-2T is a membrane-permeable RNA aptamers-activated fluorescence probe (ex/em=500 nm/523 nm). It is used to image RNA in live cells.
  • For research use only.
  • Not independently confirmed for accuracy of methods.
Catalog No. 50-003-44013 Supplier Medchemexpress LLC Supplier No. HY110251A100MG
DFHBI-2T is a membrane-permeable RNA aptamers-activated fluorescence probe (ex/em=500 nm/523 nm). It is used to image RNA in live cells.
  • For research use only.
  • Not independently confirmed for accuracy of methods.