Learn More
Aviva Systems Biology Corp DFFB Antibody - middle region (ARP52046_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52046_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DFFB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DFFB. Alternate names: CAD, CPAN, DFF-40, DFF2, DFF40ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60672