Learn More
Aviva Systems Biology Corp GSDME Antibody - C-terminal region (ARP46106_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46106_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DFNA5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DFNA5. Alternate names: ICERE-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-59227