Learn More
Apexbio Technology LLC DFO(Synonyms: 1,8-Diazafluoren-9-one, 1,8-Diazafluorenone, 9H-Pyrido[3,4-b]indol-9-one), 100mg, CAS: 54078-29-4.
SDP
Catalog No. 5000061372
Encompass_Preferred
DFO (CAS 54078-29-4) is a fluorescent dye widely utilized in forensic analyses due to its ability to chemically interact with amino acids present in latent fingerprints Upon reaction it emits fluorescence facilitating visualization and enhancing detection sensitivity particularly on porous substrates Its reactive mechanism with trace amino acid residues enables efficient fingerprint identification establishing it as an important reagent in forensic research and investigative methodologies
Catalog No. 50-000-61372 Supplier Apexbio Technology LLC Supplier No. C6997100
DFO (CAS 54078-29-4) is a fluorescent dye widely utilized in forensic analyses due to its ability to chemically interact with amino acids present in latent fingerprints Upon reaction it emits fluorescence facilitating visualization and enhancing detection sensitivity particularly on porous substrates Its reactive mechanism with trace amino acid residues enables efficient fingerprint identification establishing it as an important reagent in forensic research and investigative methodologies