Learn More
Medchemexpress LLC HY-D0903 50mg Medchemexpress, DFO CAS:54078-29-4 Purity:>98%
SDP
Catalog No. 501933207
Encompass_Preferred
Medchemexpress, HY-D0903 50mg DFO CAS:54078-29-4 DFO (9H-1,8-Diazafluoren-9-one) is a chemical that is used to find fingerprints on porous surfaces. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3207 Supplier Medchemexpress LLC Supplier No. HYD090350MG
Medchemexpress, HY-D0903 50mg DFO CAS:54078-29-4 DFO (9H-1,8-Diazafluoren-9-one) is a chemical that is used to find fingerprints on porous surfaces. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.