Learn More
TARGETMOL CHEMICALS INC DFP00173 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T110145MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DFP00173 is a potent and selective aquaporin 3 (AQP3) inhibitor. DFP00173 inhibits mouse and human AQP3 with IC50 of ~0.1-0.4 uM. Compared with the homologous AQP subtypes AQP7 and AQP9 DFP00173 is selective for AQP3. purity: 99%
Catalog No. 50-258-1600