Learn More
Medchemexpress LLC DFP00173 | 672286-03-2 | MFCD00121277 | 99.5% | 332.16 g/mol | C11H7Cl2N3O3S | 50MG
SDP
Catalog No. 5000235013
Encompass_Preferred
DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) used for biochemical and pharmacological studies. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM, is supplied as a high-purity solid, and is soluble in DMSO with defined storage recommendations.
Catalog No. 50-002-35013 Supplier Medchemexpress LLC Supplier No. HY12607350MG
DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) used for biochemical and pharmacological studies. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM, is supplied as a high-purity solid, and is soluble in DMSO with defined storage recommendations.