Learn More
Medchemexpress LLC DFRF4539A RG-7598 1mg | 1mg
SDP
Catalog No. 5000324363
Encompass_Preferred
DFRF4539A (RG-7598) is a human-derived antibody expressed in CHO that targets FcRH5/IRTA2/CD307e DFRF4539A (RG-7598) carries a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for DFRF4539A (RG-7598) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24363 Supplier Medchemexpress LLC Supplier No. HYP9905751MG
DFRF4539A (RG-7598) is a human-derived antibody expressed in CHO that targets FcRH5/IRTA2/CD307e DFRF4539A (RG-7598) carries a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for DFRF4539A (RG-7598) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)