Learn More
Medchemexpress LLC DfTat | 2035480-78-3 | 99.5% | 4076.83 | 5 MG
SDP
Catalog No. 5000396973
Encompass_Preferred
DfTat is a dimer of the prototypical cell-penetrating peptide TAT. It can efficiently deliver small molecules, peptides, and proteins into live cells. When labeled with rhodamine, DfTat can be used as a tracer for easy detection.
  • Dimer of cell-penetrating peptide TAT
  • Delivers small molecules, peptides, and proteins into live cells
  • Offers high efficiency for delivery
  • Labeled with rhodamine for easy detection
Catalog No. 50-003-96973 Supplier Medchemexpress LLC Supplier No. HYP34325MG
DfTat is a dimer of the prototypical cell-penetrating peptide TAT. It can efficiently deliver small molecules, peptides, and proteins into live cells. When labeled with rhodamine, DfTat can be used as a tracer for easy detection.
  • Dimer of cell-penetrating peptide TAT
  • Delivers small molecules, peptides, and proteins into live cells
  • Offers high efficiency for delivery
  • Labeled with rhodamine for easy detection