Learn More
Cayman Chemical DG-172hydrochlorIde 25mg
SDP
Catalog No. 502982936
Encompass_Preferred
An orally available inverse agonist of PPARβ/δ (IC50 = 27 nM), down-regulating transcription of ANGPTL4 in mouse myoblasts (IC50 = 9.5 nM)
Catalog No. 50-298-2936 Supplier Cayman Chemical Supplier No. 1912825 MG
An orally available inverse agonist of PPARβ/δ (IC50 = 27 nM), down-regulating transcription of ANGPTL4 in mouse myoblasts (IC50 = 9.5 nM)