Learn More
Cayman Chemical DG-172 (hydrochloride), Purity: ≥98%, CAS: 1361504-77-9, 5 mg
SDP
Catalog No. 502982284
Encompass_Preferred
An orally available inverse agonist of PPARβ/δ (IC50 = 27 nM), down-regulating transcription of ANGPTL4 in mouse myoblasts (IC50 = 9.5 nM)
Catalog No. 50-298-2284 Supplier Cayman Chemical Supplier No. 191285 MG
An orally available inverse agonist of PPARβ/δ (IC50 = 27 nM), down-regulating transcription of ANGPTL4 in mouse myoblasts (IC50 = 9.5 nM)