Learn More
NEOGEN Dichloran Glycerol (DG-18) Agar Base, 5 kg
SDP
Catalog No. 502015391
Encompass_Preferred
Dichloran Glycerol (DG-18) Agar Base is used for the selective isolation and enumeration of yeasts and molds from foods in a laboratory setting. Dichloran Glycerol (DG-18) Agar Base is not intended for use in the diagnosis of disease or other conditions in humans. Dichloran Glycerol (DG-18) Agar Base is used for the enumeration of viable osmophilic yeasts and xerophilic molds in food or animal feed products with a water activity of less than or equal to 0.95. This includes such foods as dry fruits, jams, cakes, dried meat, salted fish, grains, cereals, flours, nuts, spices, condiments and some animal feeds. This medium is not suitable for the examination of dehydrated products with a water activity of greater than 0.95, Dichloran Rose Bengal Chloramphenicol Agar (NCM0082 or NCM0029) should be used.For more information visit https://www.neogen.com/solutions/Microbiology/dichloran-glycerol-agar-base
Catalog No. 50-201-5391 Supplier NEOGEN Supplier No. 700003198
Dichloran Glycerol (DG-18) Agar Base is used for the selective isolation and enumeration of yeasts and molds from foods in a laboratory setting. Dichloran Glycerol (DG-18) Agar Base is not intended for use in the diagnosis of disease or other conditions in humans. Dichloran Glycerol (DG-18) Agar Base is used for the enumeration of viable osmophilic yeasts and xerophilic molds in food or animal feed products with a water activity of less than or equal to 0.95. This includes such foods as dry fruits, jams, cakes, dried meat, salted fish, grains, cereals, flours, nuts, spices, condiments and some animal feeds. This medium is not suitable for the examination of dehydrated products with a water activity of greater than 0.95, Dichloran Rose Bengal Chloramphenicol Agar (NCM0082 or NCM0029) should be used.For more information visit https://www.neogen.com/solutions/Microbiology/dichloran-glycerol-agar-base