Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DG013A (formate) | 00-00-0 | 99.6% | 530.99 g/mol | C27H37N4O4P · 2/5CH2O2 | 5 MG
SDP

Catalog No. 5000223066
Encompass_Preferred

DG013A (formate) is a phosphinic acid tripeptide mimetic inhibitor of endoplasmic reticulum aminopeptidases ERAP1 and ERAP2, used as a biochemical probe in studies of antigen processing, autoimmune disease, and cancer research. Reported biochemical potency supports use in enzyme inhibition assays and target-validation studies.

  • Phosphinic acid tripeptide mimetic inhibitor of ERAP enzymes.
  • Potent activity: ERAP1 IC50 33 nM, ERAP2 IC50 11 nM.
  • High purity by LCMS: 99.6%.
  • Molecular weight: 530.99 g/mol.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Provided for research use; not for human or clinical use.

Catalog No. 50-002-23066 Supplier Medchemexpress LLC Supplier No. HY139907B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DG013A (formate) is a phosphinic acid tripeptide mimetic inhibitor of endoplasmic reticulum aminopeptidases ERAP1 and ERAP2, used as a biochemical probe in studies of antigen processing, autoimmune disease, and cancer research. Reported biochemical potency supports use in enzyme inhibition assays and target-validation studies.

  • Phosphinic acid tripeptide mimetic inhibitor of ERAP enzymes.
  • Potent activity: ERAP1 IC50 33 nM, ERAP2 IC50 11 nM.
  • High purity by LCMS: 99.6%.
  • Molecular weight: 530.99 g/mol.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Provided for research use; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.