Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DG013A (formate) | 00-00-0 | 99.6% | C27H37N4O4P.2/5CH2O2 | 10MG
SDP

Catalog No. 5000262027
Encompass_Preferred

DG013A (formate) is a phosphinic acid tripeptide-mimetic inhibitor for research use targeting endoplasmic reticulum aminopeptidases ERAP1 and ERAP2. It is supplied as a powder with molecular formula C27H37N4O4P·2/5CH2O2 and molecular weight 530.99; reported potency values are ERAP1 IC50 = 33 nM and ERAP2 IC50 = 11 nM.

  • Phosphinic acid tripeptide-mimetic inhibitor.
  • Potent ERAP1 and ERAP2 inhibition (IC50 33 nM and 11 nM).
  • High purity (99.6% by LCMS).
  • Powder form for easy handling and storage.
  • Stable as powder at -20°C for up to 3 years.
  • In-solvent stability: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-62027 Supplier Medchemexpress LLC Supplier No. HY139907B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DG013A (formate) is a phosphinic acid tripeptide-mimetic inhibitor for research use targeting endoplasmic reticulum aminopeptidases ERAP1 and ERAP2. It is supplied as a powder with molecular formula C27H37N4O4P·2/5CH2O2 and molecular weight 530.99; reported potency values are ERAP1 IC50 = 33 nM and ERAP2 IC50 = 11 nM.

  • Phosphinic acid tripeptide-mimetic inhibitor.
  • Potent ERAP1 and ERAP2 inhibition (IC50 33 nM and 11 nM).
  • High purity (99.6% by LCMS).
  • Powder form for easy handling and storage.
  • Stable as powder at -20°C for up to 3 years.
  • In-solvent stability: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.