Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DG013A formate | 2007955-07-7 | 99.6% | 530.99 | 1 MG
SDP

Catalog No. 5000347854
Encompass_Preferred

DG013A formate is a phosphinic acid tripeptide mimetic inhibitor that displays significantly weak affinity for both ERAP1 (IC50=33 nM) and ERAP2 (IC50=11 nM). It can be used for the research of autoimmune disease and cancer.

  • Phosphinic acid tripeptide mimetic inhibitor
  • Displays weak affinity for ERAP1 (IC50=33 nM) and ERAP2 (IC50=11 nM)
  • Can be used for research of autoimmune disease and cancer

Catalog No. 50-003-47854 Supplier Medchemexpress LLC Supplier No. HY139907B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DG013A formate is a phosphinic acid tripeptide mimetic inhibitor that displays significantly weak affinity for both ERAP1 (IC50=33 nM) and ERAP2 (IC50=11 nM). It can be used for the research of autoimmune disease and cancer.

  • Phosphinic acid tripeptide mimetic inhibitor
  • Displays weak affinity for ERAP1 (IC50=33 nM) and ERAP2 (IC50=11 nM)
  • Can be used for research of autoimmune disease and cancer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.