Learn More
Medchemexpress LLC DG013B formate | 99.6% | 650.66 | C30H43N4O10P | 5 MG
SDP
Catalog No. 5000223300
Encompass_Preferred
DG013B formate is an epimeric phosphinic acid tripeptide-mimetic compound with weak affinity for ERAP1 and ERAP2. It is commonly used as a negative control in biochemical and binding studies involving aminopeptidases and related analogues.
Catalog No. 50-002-23300 Supplier Medchemexpress LLC Supplier No. HY139907C5MG
DG013B formate is an epimeric phosphinic acid tripeptide-mimetic compound with weak affinity for ERAP1 and ERAP2. It is commonly used as a negative control in biochemical and binding studies involving aminopeptidases and related analogues.