Learn More
Medchemexpress LLC HY-10825 5mg Medchemexpress, DG051 CAS:929915-58-2 Purity:>98%
SDP
Catalog No. 501968514
Encompass_Preferred
Medchemexpress, HY-10825 5mg DG051 CAS:929915-58-2 DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8514 Supplier Medchemexpress LLC Supplier No. HY108255MG
Medchemexpress, HY-10825 5mg DG051 CAS:929915-58-2 DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.