Learn More
ATCC DG-75; Burkitt's Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL2625
Encompass
DG-75 [D.G.-75] is a B lymphocyte cell line that was isolated from a pleural effusion in 1975 from a 10-year-old, White, male patient with Burkitt’s lymphoma. This cell line was deposited by H Ben-Bassat and can be used in immunology research.
Catalog No. 50-238-4803