Learn More
Eichrom Industries DGA Resin, Normal, 50 grams, 50-100 μm
SDP
Catalog No. NC1447751
Encompass
Eichrom Technologies DN-B50-S DGA Normal Resin is an extraction chromatographic material designed for analytical separations within radiochemistry. Packaged in a convenient 50-gram bottle, it features a highly selective extractant system made from N,N,N',N'-tetra-n-octyldiglycolamide. The resin's optimized particle size enhances flow and separation efficiency, making it ideal for laboratory procedures requiring precise and reliable analyte isolation.
  • Extraction chromatography resin for analytical separations
  • Packaged in a 50-gram bottle
  • Particle size of 50-100 μm ensures efficient flow
  • High selectivity extractant system based on N,N,N',N'-tetra-n-octyldiglycolamide
  • Bed density of approximately 0.38 g/mL
  • Suitable for radiochemical separations and environmental analysis
  • Hazard warnings: Harmful if swallowed, causes skin and serious eye irritation, may cause respiratory irritation
Catalog No. NC1447751 Supplier Eichrom Industries Supplier No. DNB50S
Eichrom Technologies DN-B50-S DGA Normal Resin is an extraction chromatographic material designed for analytical separations within radiochemistry. Packaged in a convenient 50-gram bottle, it features a highly selective extractant system made from N,N,N',N'-tetra-n-octyldiglycolamide. The resin's optimized particle size enhances flow and separation efficiency, making it ideal for laboratory procedures requiring precise and reliable analyte isolation.
  • Extraction chromatography resin for analytical separations
  • Packaged in a 50-gram bottle
  • Particle size of 50-100 μm ensures efficient flow
  • High selectivity extractant system based on N,N,N',N'-tetra-n-octyldiglycolamide
  • Bed density of approximately 0.38 g/mL
  • Suitable for radiochemical separations and environmental analysis
  • Hazard warnings: Harmful if swallowed, causes skin and serious eye irritation, may cause respiratory irritation