Learn More
Eichrom Industries DGA Resin, 2 mL cartridge, pack of 50, Normal DGA resin, 50-100 μm particle size, approx. 0.38 g/mL bed density, elution with 0.05-2.0M HCl
SDP
Catalog No. NC1569048
Encompass
DGA Resin, 2 mL cartridge, pack of 50, Normal DGA resin, 50-100 μm particle size, approx. 0.38 g/mL bed density, elution with 0.05-2.0M HCl
Catalog No. NC1569048 Supplier Eichrom Industries Supplier No. DNR50S
DGA Resin, 2 mL cartridge, pack of 50, Normal DGA resin, 50-100 μm particle size, approx. 0.38 g/mL bed density, elution with 0.05-2.0M HCl