Learn More
Aviva Systems Biology Corp Dgat2 Antibody - C-terminal region (ARP50084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dgat2. It was validated on Western Blot. . Alternate names: 0610010B06Rik, DGAT-2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-483