Learn More
Aviva Systems Biology Corp DGAT2L4 Antibody - C-terminal region (ARP44425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGAT2L4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DGAT2L4. Alternate names: AWAT2, DC4, MFAT, WS, DGAT2L4ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53420