Learn More
Aviva Systems Biology Corp DGAT2L7 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP49274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGAT2L7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DGAT2L7. Alternate names: DGAT2L7, ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-285-410