Learn More
Aviva Systems Biology Corp DGCR2 Antibody - middle region (ARP46576_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46576_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGCR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DGCR2. Alternate names: DGS-C, DKFZp686I1730, IDD, KIAA0163, LAN, SEZ-12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59352