Learn More
Aviva Systems Biology Corp DGCR8 Antibody - N-terminal region (ARP40984_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40984_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGCR8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DGCR8. Alternate names: C22orf12, DGCRK6, Gy1, pashaApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53255