Learn More
Proteintech Group Inc DGCR8 C-terminal Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 109961AP150UL
Encompass_Preferred
The DGCR8 C-terminal antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DGCR8 C-terminal. This antibody recognizes human, mouse, rat antigen. The DGCR8 C-terminal antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Catalog No. 50-225-6366