Learn More
Electron Microscopy Sciences DGD Embedding Kit
SDP
Supplier:  Electron Microscopy Sciences 14010
Encompass_Preferred
DGD is a removable embedding medium (for thin and thick sections) that provides embedment free sections. Good for immunolabeling and high resolution light microscopy. Section are easy to cut and ribbons are easily obtainable. Ethanol and n-butyl alcohol are used as dehydrating agents. Kit consists of: 450 ml DGD, 450 ml Ethanol, 450 ml n-Butyl alcohol, DGD Embedding Kit
Catalog No. 50-980-383