Learn More
Sino Biological Recombinant Human SIRT7 Protein (Full Length, His Tag), Active
Catalog No. 502573020
Encompass_Preferred
Recombinant full length human SIRT7 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-3020 Supplier Sino Biological Supplier No. S4130H500
Recombinant full length human SIRT7 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.