Learn More
Sino Biological Recombinant Human PLCD4 Protein (Full Length, GST Tag)
Catalog No. 502573666
Encompass_Preferred
Recombinant full length human PLCD4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3666 Supplier Sino Biological Supplier No. P44230DG1000
Add to Cart
Add to Cart
Request a Quote
Recombinant full length human PLCD4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.