Learn More
Aviva Systems Biology Corp DGKA Antibody - N-terminal region (ARP53483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGKA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DGKA. Alternate names: DAGK, DAGK1, DGK-alpha, MGC12821, MGC42356ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61036