Learn More
Sino Biological Recombinant Human DGKG Protein (Full Length, His Tag), Active
Catalog No. 502575358
Encompass_Preferred
Recombinant full length isoform 1 of human Diacylglycerol kinase gamma (DGKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5358 Supplier Sino Biological Supplier No. D2510BH500
Recombinant full length isoform 1 of human Diacylglycerol kinase gamma (DGKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.