Learn More
Aviva Systems Biology Corp DGKH Antibody - middle region (ARP45738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGKH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DGKH. Alternate names: DGKeta, DKFZp761I1510ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59123