Learn More
Cayman Chemical DGlucosamIn6phosphatehy 100mg
SDP
Catalog No. 502990245
Encompass_Preferred
A monosaccharide intermediate of the hexosamine biosynthetic pathway; binds the GlmS riboswitch (Ki = 380 µM) and inhibits GlmS production and decreases D-GlcN-6-P levels; precursor in the biosynthesis of UDP-GlcNAc
Catalog No. 50-299-0245 Supplier Cayman Chemical Supplier No. 30869100 MG
A monosaccharide intermediate of the hexosamine biosynthetic pathway; binds the GlmS riboswitch (Ki = 380 µM) and inhibits GlmS production and decreases D-GlcN-6-P levels; precursor in the biosynthesis of UDP-GlcNAc