Learn More
Cayman Chemical D-Glucosamine-6-phosphate (hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502990246
Encompass_Preferred
A monosaccharide intermediate of the hexosamine biosynthetic pathway; binds the GlmS riboswitch (Ki = 380 µM) and inhibits GlmS production and decreases D-GlcN-6-P levels; precursor in the biosynthesis of UDP-GlcNAc
Catalog No. 50-299-0246 Supplier Cayman Chemical Supplier No. 3086950 MG
A monosaccharide intermediate of the hexosamine biosynthetic pathway; binds the GlmS riboswitch (Ki = 380 µM) and inhibits GlmS production and decreases D-GlcN-6-P levels; precursor in the biosynthesis of UDP-GlcNAc