Learn More
Electron Microscopy Sciences Digital Megamag-Genie, 50hz, Australian Plug, 230 V
SDP
Catalog No. 501899877
Encompass_Preferred
The powerful single-position MegaMag Genie and Digital MegaMag Genie magnetic stirrer provide the optimal stirring for large volume containers. Powerful magnet and motor offer exceptional magnetic force capable of stirring up to 38L. Can Be Used in cold rooms and incubators.
Catalog No. 50-189-9877 Supplier Electron Microscopy Sciences Supplier No. 72401AU
The powerful single-position MegaMag Genie and Digital MegaMag Genie magnetic stirrer provide the optimal stirring for large volume containers. Powerful magnet and motor offer exceptional magnetic force capable of stirring up to 38L. Can Be Used in cold rooms and incubators.