Learn More
Cole-Parmer Shimpo DT-315A Digital Battery Powered Stroboscope
Catalog No. 50993610 Shop All Cole-Parmer Products
Encompass
The portable stroboscopes are designed with a robust, extruded aluminum housing with a repeatable accuaracy of ±0.01% of reading. Other features include a continuous duty cycle, phase shift, microprocessor, external trigger, snchronous output, flash timer control, highly polished focused reflector, and manual flash controller knob.
Catalog No. 50-993-610 Supplier Cole-Parmer Supplier No. 9880634
The portable stroboscopes are designed with a robust, extruded aluminum housing with a repeatable accuaracy of ±0.01% of reading. Other features include a continuous duty cycle, phase shift, microprocessor, external trigger, snchronous output, flash timer control, highly polished focused reflector, and manual flash controller knob.