Learn More
Bioworld dGTP, 100mM Solution, 40 μmol
Catalog No. 50255850
Encompass_Preferred
Ultra Pure Grade2'-Deoxyguanosine-5'-triphosphateStability (-20°C): >1yearMax wavelength (nm) : 250-254 Appearance: LiquidPH Value: pH: 7.0 ± 0.5
Catalog No. 50-255-850 Supplier Bioworld Supplier No. 404000481
Ultra Pure Grade2'-Deoxyguanosine-5'-triphosphateStability (-20°C): >1yearMax wavelength (nm) : 250-254 Appearance: LiquidPH Value: pH: 7.0 ± 0.5