Learn More
Applied Biosystems™ dGTP BigDye™ Terminator Ready Reaction Kit
Catalog No. 4307176
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100
Catalog No. No. of Reactions
43-071-76 100
1 options
Catalog No. 43-071-76 Supplier Applied Biosystems™ Supplier No. 4307176
Add to Cart
Add to Cart
Ideal for working with GT- and G-rich and other difficult-to-sequence templates
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Description dGTP BigDye™ Terminator Ready Reaction Kit
No. of Reactions 100
Product Type dGTP BigDye™ Terminator Ready Reaction Kit