Learn More
Sigma Aldrich Fine Chemicals Biosciences dGTP PCR Grade sodium salt
Catalog No. 502903452
Encompass
dGTP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).
Catalog No. 50-290-3452 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11969030001
dGTP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).