Learn More
New England Biolabs, Inc. dGTP Solution - 25 umol
SDP
Catalog No. 502023998
Encompass
dGTP Solution contains 0.25 ml of 100 mM ultrapure dGTP as a sodium salt at pH 7.5. It is supplied in Ultrapure water as a sodium salt at pH 7.5. The dGTP Solution is supplied at a concentration of 100 mM (+/- 5 mM) as determined by A260 absorbance. Can be diluted using sterile distilled water, preferably Milli-Q water or can be diluted using sterile TE (10 mM Tris-HCl, 1 mM EDTA (pH 7.5)).
Catalog No. 50-202-3998 Supplier New England Biolabs, Inc. Supplier No. N0442S
dGTP Solution contains 0.25 ml of 100 mM ultrapure dGTP as a sodium salt at pH 7.5. It is supplied in Ultrapure water as a sodium salt at pH 7.5. The dGTP Solution is supplied at a concentration of 100 mM (+/- 5 mM) as determined by A260 absorbance. Can be diluted using sterile distilled water, preferably Milli-Q water or can be diluted using sterile TE (10 mM Tris-HCl, 1 mM EDTA (pH 7.5)).