Learn More
Cayman Chemical Tiliroside, Purity: ≥98%, CAS: 20316-62-5, 50 mg
SDP
Catalog No. 502950251
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits CAVII (Ki = 4.6 nM); scavenges DPPH and superoxide radicals (IC50s = 12.8 and 42 µM, respectively); cytotoxic to Jurkat, HepG2, and COLO 205 cells (IC50s = 11.6, 14.3, and 55.4 µM, respectively); reduces TPA-induced ear edema (ID50 = 0.357 mg/ear); decreases triglyceride accumulation in the liver and skeletal muscle in high-fat diet-fed KKAy diabetic mice at 100 mg/kg
Catalog No. 50-295-0251 Supplier Cayman Chemical Supplier No. 3449350 MG
A flavonoid glycoside with diverse biological activities; inhibits CAVII (Ki = 4.6 nM); scavenges DPPH and superoxide radicals (IC50s = 12.8 and 42 µM, respectively); cytotoxic to Jurkat, HepG2, and COLO 205 cells (IC50s = 11.6, 14.3, and 55.4 µM, respectively); reduces TPA-induced ear edema (ID50 = 0.357 mg/ear); decreases triglyceride accumulation in the liver and skeletal muscle in high-fat diet-fed KKAy diabetic mice at 100 mg/kg