Learn More
Medchemexpress LLC DGY-06-116 | 2556836-50-9 | 99.5% | C32H33ClN8O2 | 10MG
SDP
Catalog No. 5000261237
Encompass_Preferred
DGY-06-116 is an irreversible covalent Src kinase inhibitor with potent biochemical activity (IC50 3 nM) and limited activity against FGFR1 (IC50 ~8,340 nM). It is supplied as a high-purity solid and is available as a ready-to-use DMSO solution or multiple milligram pack sizes for preclinical research.
  • Irreversible covalent Src inhibitor with IC50 3 nM.
  • Low activity against FGFR1 (IC50 ~8,340 nM).
  • High purity (99.54%).
  • Supplied as a white to off-white solid.
  • Soluble in DMSO (250 mg/mL) and compatible with in vivo formulations.
  • Stable as powder at -20°C and in solvent at -80°C.
  • Available in multiple pack sizes and as a 10 mM DMSO solution.
Catalog No. 50-002-61237 Supplier Medchemexpress LLC Supplier No. HY13660510MG
DGY-06-116 is an irreversible covalent Src kinase inhibitor with potent biochemical activity (IC50 3 nM) and limited activity against FGFR1 (IC50 ~8,340 nM). It is supplied as a high-purity solid and is available as a ready-to-use DMSO solution or multiple milligram pack sizes for preclinical research.
  • Irreversible covalent Src inhibitor with IC50 3 nM.
  • Low activity against FGFR1 (IC50 ~8,340 nM).
  • High purity (99.54%).
  • Supplied as a white to off-white solid.
  • Soluble in DMSO (250 mg/mL) and compatible with in vivo formulations.
  • Stable as powder at -20°C and in solvent at -80°C.
  • Available in multiple pack sizes and as a 10 mM DMSO solution.